Boron trichloride 1 mol/l in dichloromethane, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
176681000 176688000
176681000.EA 93.1 EUR
176681000. 176688000.
Boron trichloride 1 mol/l in dichloromethane, AcroSeal®
Boron trichloride
Formula: BCl₃
MW: 117.17 g/mol
Boiling Pt: 12.4
Melting Pt: -107
Density: 1.876
Storage Temperature: Refrigerator
MDL Number: MFCD00011313
CAS Number: 10294-34-5
EINECS: 233-658-4
UN: 2922
ADR: 8 II
Merck Index: 12,01378
Order Now
Specification Test Results
Appearance Clear colorless to brown Solution (May darken on storage)
Titration Argentometric 0.9 to 1.1 M
Titration ≥98.5 %
Chloride (Cl) ≤0.1 %
Sulfate (SO4) ≤0.07 %
Insoluble matter ≤0.01 %