Boron trichloride 1 mol/l in dichloromethane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
178934-100ML
178934-800ML
SIAL178934-100MLEA
98.7
EUR
SIAL178934-100ML
SIAL178934-800ML
Boron trichloride 1 mol/l in dichloromethane, Sigma-Aldrich®
Boron trichloride
Formula:
BCl₃ MW: 117.17 g/mol Boiling Pt: 12.4 Melting Pt: -107 Density: 1.876 Storage Temperature: Ambient |
MDL Number:
MFCD00011313 CAS Number: 10294-34-5 EINECS: 233-658-4 UN: 2927 ADR: 6.1 II Merck Index: 12,01378 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...