Boron trichloride 1 M in hexane, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
198921000 198928000
198921000.EA 77.7 EUR
198921000. 198928000.
Boron trichloride 1 M in hexane, AcroSeal®
Boron trichloride
Formula: BCl₃
MW: 117.17 g/mol
Boiling Pt: 12.4
Melting Pt: -107
Density: 0.738 g/cm³
Flash Pt: –17 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011313
CAS Number: 10294-34-5
EINECS: 233-658-4
UN: 3286
ADR: 3 I
Merck Index: 12,01378
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Solution
Titration Argentometric 0.9 to 1.1 M