Dithiodiglycol (bis(2-hydroxyethyl) disulphide), Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Hydroxyethyl disulphide, 2,2'-Dithiobisethanol, 2,2'-Dithiodiethanol, Bis(2-hydroxyethyl) disulphide
M-Clarity™ quality level = MQ200
380474-250ML 380474-50ML
SIAL380474-250MLEA 355 EUR
SIAL380474-250ML SIAL380474-50ML
Dithiodiglycol (bis(2-hydroxyethyl) disulphide), Sigma-Aldrich®
Dithiodiglycol (Bis(2-hydroxyethyl) disulphide)
Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Boiling Pt: 307
Melting Pt: 27.5
Density: 1.27 g/cm³ (20 °C)
MDL Number: MFCD00002906
CAS Number: 1892-29-1
EINECS: 217-576-6
UN: 2810
ADR: 6.1 III