Dichloroacetyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
D55008-100G D55008-500G
SIALD55008-100GEA 61.7 EUR
SIALD55008-100G SIALD55008-500G
Dichloroacetyl chloride, Sigma-Aldrich®
Dichloroacetyl chloride
Formula: C₂HCl₃O
Boiling Pt: 107 °C (1013 hPa)
Density: 1.53 g/cm³ (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00000840
CAS Number: 79-36-7
EINECS: 201-199-9
UN: 1765
ADR: 8,II
Merck Index: 13,03078