Allyl hexanoate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Hexanoic acid allyl ester
M-Clarity™ quality level = MQ300
W203203-1KG-K W203203-20KG-K W203203-4KG-K
SIALW203203-1KG-KEA 77.9 EUR
SIALW203203-1KG-K SIALW203203-20KG-K SIALW203203-4KG-K
Allyl hexanoate, Sigma-Aldrich®
Allyl hexanoate
Formula: C₉H₁₆O₂
Boiling Pt: 75 °C (1013 hPa)
Density: 0.8886 g/cm³ (20 °C)
MDL Number: MFCD00038339
CAS Number: 123-68-2
EINECS: 204-642-4