Allyl hexanoate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Hexanoic acid allyl ester
M-Clarity™ quality level = MQ300
W203211-100G-K W203211-1KG-K W203211-25G-K W203211-SAMPLE
SIALW203211-100G-KEA 152 EUR
SIALW203211-100G-K SIALW203211-1KG-K SIALW203211-25G-K SIALW203211-SAMPLE
Allyl hexanoate, Sigma-Aldrich®
Allyl hexanoate
Formula: C₉H₁₆O₂
Boiling Pt: 75 °C (1013 hPa)
Density: 0.8886 g/cm³ (20 °C)
MDL Number: MFCD00038339
CAS Number: 123-68-2
EINECS: 204-642-4