Polyethylene glycol 200, Sigma-Aldrich®
Supplier: Merck
Synonyms: PEG 200, Ethylene glycol polymer average Mn 200
M-Clarity™ quality level = MQ200
P3015-1KG P3015-20KG P3015-250G P3015-500G
SIALP3015-1KGEA 61.5 EUR
SIALP3015-1KG SIALP3015-20KG SIALP3015-250G SIALP3015-500G
Polyethylene glycol 200, Sigma-Aldrich®
Polyethylene glycol 200
Formula: H(OCH₂CH₂)nOH
MW: 200 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: –55…–40 °C
Density: 1.124 g/cm³ (20 °C)
Flash Pt: 180
Storage Temperature: Ambient
MDL Number: MFCD01779596
CAS Number: 25322-68-3
EINECS: 500-038-2