Polyethylene glycol 3,350, Sigma-Aldrich®
Supplier: Merck
Synonyms:
PEG 3,350, PEG 3350
M-Clarity™ quality level = MQ200
P4338-1KG
P4338-2KG
P4338-500G
SIALP4338-1KGEA
361
EUR
SIALP4338-1KG
SIALP4338-2KG
SIALP4338-500G
Polyethylene glycol 3,350, Sigma-Aldrich®
Polyethylene glycol 3,350
Formula:
H(OCH₂CH₂)nOH MW: 3350 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...