Polypropylene glycol, M.W. 1,000, Sigma-Aldrich®
Supplier: Merck
Synonyms: Poly(propylene glycol), Poly(propylene oxide), PPG
M-Clarity™ quality level = MQ200
202320-250G 202320-500G 202320-5G
SIAL202320-250GEA 98.6 EUR
SIAL202320-250G SIAL202320-500G SIAL202320-5G
Polypropylene glycol, M.W. 1,000, Sigma-Aldrich®
Polypropylene glycol
Formula: H(OCH₂CH₂CH₂)nOH
Storage Temperature: Ambient
MDL Number: MFCD00084446
CAS Number: 25322-69-4
EINECS: 500-039-8