Polypropylene glycol, M.W. 2,000
Supplier: Thermo Fisher Scientific
Synonyms: Poly(propylene glycol), Poly(propylene oxide), PPG
L14699.AE L14699.AP L14699.PG
L14699.AEEA 38.8 EUR
L14699.AE L14699.AP L14699.PG
Polypropylene glycol, M.W. 2,000
Polypropylene glycol
CAS No.: 25322-69-4 Molecular Formula: H[OCH(CH3)CH2]nOH Flash Point: 194°(381°F) Refractive Index: 1.4510 MDL No.: MFCD01779687
Formula: H(OCH₂CH₂CH₂)nOH
MW: 1025
Density: 1.002
Flash Pt: 194 °C (381 °F)
Storage Temperature: Ambient
MDL Number: MFCD00084446
CAS Number: 25322-69-4
EINECS: 500-039-8