Polypropylene glycol, M.W. 400
Supplier: Thermo Fisher Scientific
Synonyms: Poly(propylene glycol), Poly(propylene oxide), PPG
40811.18 40811.30
40811.18EA 39.2 EUR
40811.18 40811.30
Polypropylene glycol, M.W. 400
Polypropylene glycol
CAS No.: 25322-69-4 Molecular Formula: H[OCH(CH3)CH2]nOH Physical Form: Liquid MDL No.: MFCD01779680
Formula: H(OCH₂CH₂CH₂)nOH
Storage Temperature: Ambient
MDL Number: MFCD00084446
CAS Number: 25322-69-4
EINECS: 500-039-8