Polypropylene glycol, M.W. ∼ 425, Sigma-Aldrich®
Supplier: Merck
Synonyms: Poly(propylene glycol), Poly(propylene oxide), PPG
M-Clarity™ quality level = MQ200
202304-250G 202304-500G 202304-5G
SIAL202304-250GEA 1380 EUR
SIAL202304-250G SIAL202304-500G SIAL202304-5G
Polypropylene glycol, M.W. ∼ 425, Sigma-Aldrich®
Polypropylene glycol
Formula: H(OCH₂CH₂CH₂)nOH
Melting Pt: –50 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00084446
CAS Number: 25322-69-4
EINECS: 500-039-8