trans-Anethole, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(E)-1-Methoxy-4-(prop-1-en-1-yl)benzene
M-Clarity™ quality level = MQ300
W208604-10KG-K
W208604-1KG-K
W208604-5KG-K
SIALW208604-10KG-KEA
712
EUR
SIALW208604-10KG-K
SIALW208604-1KG-K
SIALW208604-5KG-K
trans-Anethole, Sigma-Aldrich®
trans-Anethole
Formula:
C₁₀H₁₂O Boiling Pt: 236 °C (1013 hPa) Melting Pt: 21…23 °C Density: 0.9883 g/cm³ (20 °C) Flash Pt: 96 °C |
MDL Number:
MFCD00009284 CAS Number: 4180-23-8 EINECS: 224-052-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...