ESD Safety Shoes, Lace-up, Low, SL 605 XP
Supplier: ATLAS - the shoe company
4930037/W12 4930036/W12 4930038/W12 4930039/W12 4930040/W12 4930041/W12 4930042/W12 4930043/W12 4930044/W12 4930045/W12 4930046/W12 4930047/W12 4930048/W12 4930049/W12
ATLA4930037/W12EA 0 EUR
ATLA4930037/W12 ATLA4930036/W12 ATLA4930038/W12 ATLA4930039/W12 ATLA4930040/W12 ATLA4930041/W12 ATLA4930042/W12 ATLA4930043/W12 ATLA4930044/W12 ATLA4930045/W12 ATLA4930046/W12 ATLA4930047/W12 ATLA4930048/W12 ATLA4930049/W12
ESD Safety Shoes, Lace-up, Low, SL 605 XP
Shoes and Boots
ESD safety shoes, available in multiple color combinations.
  • XP® metal free puncture-resistance
  • Alu-tec aluminium toecap
  • 3D-cushioning system
  • SOFT nubuck leather
  • Aktiv-X functional lining
  • Shoe sole: MPU® INNOFLEX system
  • Adapted for orthopedic insoles
The aktiv-X functional lining stands for the highest quality, innovative technology and offers the wearer permanent, active moisture regulation. The aktiv-X functional lining has a moisture-regulating effect and ensures that the shoes dry again quickly. It does not stain, is skin-friendly and abrasion-resistant. It is also tested according to Oeko-Tex Standard 100.
The MPU REBOUND SYSTEM enables perceptibly economical and fatigue-free walking. The basis for this excellent walking comfort is formed by the three basic components: Pressure relief, cushioning, dynamics. This is ensured by the perfect interaction of a pressure-relieving insole and the light and highly reactive MPU sole technology.
The ATLAS SOFT nubuck leather has to withstand special challenges throughout the working day. The approx. 2 mm thick full-grain cowhide is particularly characterised by its breathability and its soft, comfortable feel when worn.
Colour: Blue
Certifications: EN ISO 20345 S3 SRC.