Hematin (from porcine), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
Hematin porcine has been used as a:
polymerase chain reaction (PCR) inhibitor to evaluate its effect on the PowerQuant System used for detecting degraded DNA in samples
supplement in Haemophilus test medium to culture Haemophilus influenzae isolates for resistance studies
standard to determine the heme coefficient as a part of heme iron analysis using raw beef lung
Formula:
C₃₄H₃₃FeN₄O₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD00011615 CAS Number: 15489-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...