L-Phenylalanine, Sigma-Aldrich®
Supplier: Merck
Synonyms: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine
M-Clarity™ quality level = MQ300
W358509-1KG-K W358509-5KG-K
SIALW358509-1KG-KEA 117 EUR
SIALW358509-1KG-K SIALW358509-5KG-K
L-Phenylalanine, Sigma-Aldrich®
L-α-Phenylalanine
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 313
Melting Pt: 173
Density: 0.754 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064227
CAS Number: 63-91-2
EINECS: 200-568-1
Merck Index: 14,07271