L-Alanine

Supplier: Roth Carl

Synonyms: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine

3076.1 3076.2
ROTH3076.1EA 0 EUR
ROTH3076.1 ROTH3076.2
L-Alanine
L-Alanine
Formula: C₃H₇NO₂
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 79 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064410
CAS Number: 56-41-7
EINECS: 200-273-8
Merck Index: 13,00203

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR