Cholesterol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol
M-Clarity™ quality level = MQ200
C8503-100G C8503-1KG C8503-25G C8503-500G C8503-5KG
SIALC8503-100GEA 116 EUR
SIALC8503-100G SIALC8503-1KG SIALC8503-25G SIALC8503-500G SIALC8503-5KG
Cholesterol, Sigma-Aldrich®
Cholesterol
Formula: C₂₇H₄₆O
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 147…150 °C
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00003646
CAS Number: 57-88-5
EINECS: 200-353-2
Merck Index: 13,02221