5α-Cholestan-3-one, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

C8128-1G C8128-25G
SIALC8128-1GEA 128 EUR
SIALC8128-1G SIALC8128-25G
5α-Cholestan-3-one, Sigma-Aldrich®
5α-Cholestan-3-one
Formula: C₂₇H₄₆O
MDL Number: MFCD00065901
CAS Number: 566-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR