Heptafluorobutyric anhydride (HFAA) ≥98%, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
HFBA, 2,2,3,3,4,4,4-Heptafluorobutanoic anhydride, Perfluorobutyric anhydride, HFAA
H1006-10AMP
H1006-25ML
H1006-5ML
SIALH1006-10AMPEA
178
EUR
SIALH1006-10AMP
SIALH1006-25ML
SIALH1006-5ML
Heptafluorobutyric anhydride (HFAA) ≥98%, Sigma-Aldrich®
Heptafluorobutyric anhydride
Formula:
C₈F₁₄O₃ Boiling Pt: 108-110 °C (lit.) Melting Pt: −43 °C (lit.) Density: 1.674 g/mL (20 °C) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00000432 CAS Number: 336-59-4 EINECS: 206-410-8 UN: 3625 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...