Phenoxyacetyl chloride ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
A13761.18
A13761.30
A13761.18EA
68.3
EUR
A13761.18
A13761.30
Phenoxyacetyl chloride ≥98%
Phenoxyacetyl chloride
Purity: 98%
CAS No.: 701-99-5
Molecular Formula: C8H7ClO2
Formula Weight: 170.60
Flash Point: 108°(226°F)
Refractive Index: 1.5340
Odour: Irritating
Boiling Point: 225-226°
Beilstein Reference: 607585
UN No.: 3265
MDL No.: MFCD00000726
Formula:
C₈H₇ClO₂ Boiling Pt: 225…226 °C Melting Pt: 28…30 °C Density: 1.235 Flash Pt: 108 °C (226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000726 CAS Number: 701-99-5 EINECS: 211-862-4 UN: 3265 ADR: 8,II |