Phenoxyacetyl chloride 98%
Supplier: Thermo Fisher Scientific
Danger
169940500 169942500 169940100
ACRO169940500EA 78.3 EUR
ACRO169940500 ACRO169942500 ACRO169940100
Phenoxyacetyl chloride 98%
Phenoxyacetyl chloride
Formula: C₈H₇ClO₂
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: 28…30 °C
Density: 1.23 g/cm³
Flash Pt: 108 °C
MDL Number: MFCD00000726
CAS Number: 701-99-5
EINECS: 211-862-4
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear light yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5324 to 1.5344 (20°C, 589 nm)
Titration RCOCl ≥97.5 %