Anti-Isotype Control Mouse Monoclonal Antibody [clone: IGG1/1331] (CF®405L)

Supplier: Biotium
BNC061331-250 BNC061331-1ML
BNC061331-250EA 80.2 EUR
BNC061331-250 BNC061331-1ML
Anti-Isotype Control Mouse Monoclonal Antibody [clone: IGG1/1331] (CF®405L)
Antibodies
Non-reactive antibody to use as a negative control for mouse IgG1 primary antibodies. Available with CF® Dyes and other labels.

  • Isotype control for antibody-based experiments
  • Available with bright and photostable CF® Dyes
  • APC, R-PE, RPE-Astral™616, APC-Astral™813, and biotin conjugates also available
  • Suitable for western, flow cytometry, ELISA, immunofluorescence, and immunohistology

This is a non-reactive mouse monoclonal IgG1 kappa antibody labeled with one of our bright and photostable CF® Dyes and other labels. This antibody may serve as a control for a mouse monoclonal IgG1 antibody under identical experimental conditions. Biotium also carries mouse monoclonal isotype control antibodies for IgG2a Kappa (MG2a-53) and IgG2b Kappa (IGG1/1333).

Type: Isotype control
Antigen:
Clonality: Monoclonal
Clone:
Conjugation: CF®405L
Epitope:
Host: Mouse
Isotype: IgG1, Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR