Linoleamide ≥98% (by TLC), white solid
Supplier: ENZO LIFE SCIENCES
BMLFA0230010 BMLFA0230050
ENZOBMLFA0230010EA 66 EUR
ENZOBMLFA0230010 ENZOBMLFA0230050
Linoleamide ≥98% (by TLC), white solid
Linoleamide
Unsaturated analog of endogenous sleep inducing lipid.
Soluble in 100% ethanol or in DMSO.
Formula: C₁₈H₃₃NO
MDL Number: MFCD00798239
CAS Number: 3072-13-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f845a75711e1331b25bf57ccf753cfb9c06637e&vid=10c0afa086bbcef33061ea3c0d6869ffee6be9bc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity ≥98% (TLC)