Adenosine-5'-O-(3-thiotriphosphate) tetralithium salt ≥85% (by HPLC) ATP analog
Supplier: ENZO LIFE SCIENCES
Warning
Synonyms: Ado-5′-PPP[S], ATPγ-S . 4Li, Adenosine 5′-[γ-thio]triphosphate tetralithium salt, Adenosine 5′-(3-thiotriphosphate) tetralithium salt
BMLST1010005 BMLST1010025
ENZOBMLST1010005EA 165 EUR
ENZOBMLST1010005 ENZOBMLST1010025
Adenosine-5'-O-(3-thiotriphosphate) tetralithium salt ≥85% (by HPLC) ATP analog
Adenosine-5'-O-(3-thiotriphosphate) tetralithium salt
An ATP analog that can substitute for ATP in various kinase reactions. The resulting thiophosphorylated proteins are resistant to dephosphorylation by protein phosphatases.
Appearance: White to off-white solid
Solubility: Soluble in water (25mg/ml)
Shipping: Ambient
Long Term Storage: -20°C
Formula: C₁₀H₁₂N₅O₁₂PS·4Li
MW: 546.98 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077293
CAS Number: 93839-89-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5aefc86e47a821f4aecb306f287e2cc0fffe018a&vid=5b9784995362d84c1dbfd5571921bf83f7cc0136&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity ≥85% (HPLC)