Staurosporine ≥98% (by HPLC)
Supplier: ENZO LIFE SCIENCES
Synonyms: (5S,6R,7R,9R)-6-Methoxy-5-methyl-7-(methylamino)-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-14-one, Antibiotic AM-2282
ALX380014C250 ALX380014M001 ALX380014M005
ENZOALX380014C250EA 165 EUR
ENZOALX380014C250 ENZOALX380014M001 ENZOALX380014M005
Staurosporine ≥98% (by HPLC)
Staurosporine
Apoptosis inducer.
  • Protein kinase inhibitor
  • Isolated from Streptomyces staurosporeus
  • Off-white to green powder
  • Soluble in ethyl acetate, DMSO (25 mg/ml); dimethyl formamide (25 mg/ml) and slightly soluble in chloroform and methanol
  • Insoluble in water
Potent cell-permeable inhibitor of a variety of protein kinases, e.g. protein kinase C (PKC), CDK1/cyclin B (IC50~5 nM), CDK2/cyclin A (IC50=7 nM), CDK4/cyclin D (IC50=3-10 µM), CDK5/p25 (IC50=4 nM), GSK-3β (IC50=15 nM), Pim-1 kinase (IC50=10 nM).Binds do the ATP binding domain. Did not inhibit PKC-ζ. At 1 µM induced apoptosis in CHO cells. Inhibits topoisomerase II directly by blocking transfer of phosphodiester bonds from DNA to active site tyrosine.
Formula: C₂₈H₂₆N₄O₃
MW: 466.541 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18252446
CAS Number: 62996-74-1