Concanamycin A (from Streptomyces species) ≥98% (by HPLC)
Supplier: ENZO LIFE SCIENCES
ALX380034C025 ALX380034C100 ALX380034M001
ENZOALX380034C025EA 260 EUR
ENZOALX380034C025 ENZOALX380034C100 ENZOALX380034M001
Concanamycin A (from Streptomyces species) ≥98% (by HPLC)
Concanamycin A
White to off-white solid (sub-milligram quantities will appear as clear film)
Antibiotic. More potent and specific H+-ATPase inhibitor than bafilomycin A1. Inhibits acidification of organelles such as lysosomes and the Golgi apparatus. Blocks cell surface expression of viral glycoproteins without affecting their synthesis. Exhibits cytotoxic effects in a number of cell lines in a cell viability assay. Induces nitric oxide (NO) production. Inhibits Autophagy.
Soluble in methanol, DMSO or acetonitrile, insoluble in water
Packaging: Shipping temperature: Ambient
Formula: C₄₆H₇₅NO₁₄
MW: 866.1 g/mol
Storage Temperature: Freezer
CAS Number: 80890-47-7