Colchicine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
7α-H-Colchicine, (S)-(-)-Colchicine
M-Clarity™ quality level = MQ200
C3915-1G
C3915-500MG
C3915-5G
SIALC3915-1GEA
251
EUR
SIALC3915-1G
SIALC3915-500MG
SIALC3915-5G
Colchicine, Sigma-Aldrich®
Colchicine
Formula:
C₂₂H₂₅NO₆ MW: 399.44 g/mol Melting Pt: 154…156 °C Density: 1.32 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00078484 CAS Number: 64-86-8 EINECS: 200-598-5 UN: 1544 ADR: 6.1 I Merck Index: 12,02536 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...