4-Chlorostyrene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

C71203-10G C71203-50G
SIALC71203-10GEA 75.9 EUR
SIALC71203-10G SIALC71203-50G
4-Chlorostyrene, Sigma-Aldrich®
4-Chlorostyrene
Formula: C₈H₇Cl
Boiling Pt: 192 °C (1013 hPa)
Melting Pt: –15.9 °C
Density: 1.155 g/cm³ (20 °C)
MDL Number: MFCD00000632
CAS Number: 1073-67-2
EINECS: 214-028-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR