Methyl red sodium salt, Sigma-Aldrich®
Supplier: Merck
Synonyms: Methyl red water soluble, 2-(4-Dimethylaminophenylazo)benzoic acid sodium salt, Sodium 2-(p-(dimethylamino)phenylazo)benzoate
M-Clarity™ quality level = MQ200
114502-100G 114502-25G
SIAL114502-100GEA 145 EUR
SIAL114502-100G SIAL114502-25G
Methyl red sodium salt, Sigma-Aldrich®
Methyl red sodium salt
Formula: C₁₅H₁₄N₃NaO₂
MW: 291.28 g/mol
Melting Pt: > 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00002426
CAS Number: 845-10-3
EINECS: 212-682-9
Merck Index: 13,06142