Methyl red sodium salt 0,02% (w/v) in aqueous solution
Supplier: Thermo Fisher Scientific
Synonyms: Methyl red water soluble, 2-(4-Dimethylaminophenylazo)benzoic acid sodium salt, Sodium 2-(p-(dimethylamino)phenylazo)benzoate
44161.AE 44161.AP
44161.AEEA 27.9 EUR
44161.AE 44161.AP
Methyl red sodium salt 0,02% (w/v) in aqueous solution
Methyl red sodium salt
CAS No.: 845-10-3 Molecular Formula: (CH3)2NC6H4N=NC6H4CO2Na Formula Weight: 291.29 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00002426
Formula: C₁₅H₁₄N₃NaO₂
MW: 291.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002426
CAS Number: 845-10-3
EINECS: 212-682-9
Merck Index: 13,06142