2-Amino-6,7-dimethyl-4-hydroxypteridine

Supplier: Thermo Scientific

RJC00673.1 RJC00673.10
MAYBRJC00673.1EA 0 EUR
MAYBRJC00673.1 MAYBRJC00673.10
2-Amino-6,7-dimethyl-4-hydroxypteridine
2-Amino-6,7-dimethyl-4-hydroxypteridine
Formula: C₈H₉N₅O
CAS Number: 611-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR