Bromothymol blue sodium salt, Sigma-Aldrich®
Supplier: Merck
Synonyms: Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulphonate, Bromothymol blue water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-isopropyl-3-methylphenolate
18470-10G 18470-1G 18470-50G
SIAL18470-10GEA 62.7 EUR
SIAL18470-10G SIAL18470-1G SIAL18470-50G
Bromothymol blue sodium salt, Sigma-Aldrich®
Bromothymol blue sodium salt
Formula: C₂₇H₂₇Br₂NaO₅S
Melting Pt: > 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00077263
CAS Number: 34722-90-2
EINECS: 252-169-7