4-(Dimethylamino)benzaldehyde
Supplier: Roth Carl
X867.1 X867.2
ROTHX867.1EA 56.6 EUR
ROTHX867.1 ROTHX867.2
4-(Dimethylamino)benzaldehyde
4-(Dimethylamino)benzaldehyde
Formula: (CH₃)₂NC₆H₄CHO
Boiling Pt: 176…177 °C (1013 hPa)
Melting Pt: 72 °C
Density: 1.0254 g/cm³ (99.9 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS Number: 100-10-7
EINECS: 202-819-0
Merck Index: 13,03257
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=909765f6b48eb3673cc290753e0ead3358b739cf&prid=web&clid=SAPDAM