Sodium alginate, low viscosity
Supplier: Thermo Fisher Scientific
Synonyms: Alginic acid sodium salt, Algin
B25266.22 B25266.0E B25266.36
B25266.22EA 53.6 EUR
B25266.22 B25266.0E B25266.36
Sodium alginate, low viscosity
Sodium alginate
CAS No.: 9005-38-3 Merck Reference: 14,241 MDL No.: MFCD00081310
Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3
Merck Index: 14,241