Sodium alginate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Alginic acid sodium salt, Algin
M-Clarity™ quality level = MQ300
W201502-1KG W201502-5KG W201502-SAMPLE
SIALW201502-1KGEA 145 EUR
SIALW201502-1KG SIALW201502-5KG SIALW201502-SAMPLE
Sodium alginate, Sigma-Aldrich®
Sodium alginate
Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3