Colophony (Rosin gum)
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Rosin, gum, Rosin, Gum rosin
36734.A1 36734.30
36734.A1EA 105 EUR
36734.A1 36734.30
Colophony (Rosin gum)
Colophony (Rosin gum)
CAS No.: 8050-09-7 Physical Form: Powder/solid Solubility: Soluble in alcohol, benzene, ether. Insoluble in water Merck Reference: 14,8266 MDL No.: MFCD00132205
Melting Pt: 75…81 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00132205
CAS Number: 8050-09-7
EINECS: 232-475-7
Merck Index: 14,8266