Colophony (Rosin gum), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Rosin, gum, Rosin, Gum rosin
60895-1KG 60895-250G
SIAL60895-1KGEA 165 EUR
SIAL60895-1KG SIAL60895-250G
Colophony (Rosin gum), Sigma-Aldrich®
Colophony (Rosin gum)
Melting Pt: 75…81 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 188 °C
MDL Number: MFCD00132205
CAS Number: 8050-09-7
EINECS: 232-475-7