Sodium stannate trihydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Sodium tin(IV) oxide trihydrate, Disodium tin trioxide trihydrate
MQ200
336262-100G 336262-500G
SIAL336262-100GEA 40.3 EUR
SIAL336262-100G SIAL336262-500G
Sodium stannate trihydrate, Sigma-Aldrich®
Sodium stannate trihydrate
Formula: Na₂O₃Sn·3H₂O
MW: 266.73 g/mol
MDL Number: MFCD00150189
CAS Number: 12209-98-2
EINECS: 235-030-5