Turpentine oil, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Oil of turpentine, Fir oil, Turpentine, Turpentine, Rectified, Pine oil, L-Turpentine
M-Clarity™ quality level = MQ100
24245-2.5L 24245-1L
SIAL24245-2.5LEA 146 EUR
SIAL24245-2.5L SIAL24245-1L
Turpentine oil, Sigma-Aldrich®
Turpentine oil
Boiling Pt: 153…175 °C (1013 hPa)
Melting Pt: –60…–50 °C
Density: 0.854…0.868 g/cm³ (25 °C)
Flash Pt: 35 °C
MDL Number: MFCD00147790
CAS Number: 8006-64-2
EINECS: 232-350-7
UN: 1299
ADR: 3,III