Turpentine oil

Supplier: Roth Carl
Danger

Synonyms: Oil of turpentine, Fir oil, Turpentine, Turpentine, Rectified, Pine oil, L-Turpentine

T139.1 T139.2 T139.3
ROTHT139.1EA 86.5 EUR
ROTHT139.1 ROTHT139.2 ROTHT139.3
Turpentine oil
Turpentine oil
Boiling Pt: 153…175 °C (1013 hPa)
Melting Pt: –60…–50 °C
Density: 0.854…0.868 g/cm³ (25 °C)
Flash Pt: 35 °C
MDL Number: MFCD00147790
CAS Number: 8006-64-2
EINECS: 232-350-7
UN: 1299
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR