Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (mFluor™ Green 620)

Supplier: AAT BIOQUEST
100320U0 100320U1
AATB100320U0EA 371 EUR
AATB100320U0 AATB100320U1
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (mFluor™ Green 620)
Antibodies
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kDa member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependent)s.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: mFluor™ Green 620
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR