Anti-CD16, alpha chain Mouse Monoclonal Antibody [clone: 3G8] (mFluor™ Violet 500)

Supplier: AAT BIOQUEST
10161100 10161101
AATB10161100EA 371 EUR
AATB10161100 AATB10161101
Anti-CD16, alpha chain Mouse Monoclonal Antibody [clone: 3G8] (mFluor™ Violet 500)
Antibodies
The 3G8 monoclonal antibody recognises human CD16a, a 50 to 65 kD transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.

  • Affinity purified
  • Easily reconstituted

Caution: For research use only.

Type: Primary
Antigen: CD16 antigen, alpha chain
Clonality: Monoclonal
Clone: 3G8
Conjugation: mFluor™ Violet 500
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR