Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (mFluor™ Violet 540)

Supplier: AAT BIOQUEST
10041120 10041121
AATB10041120EA 371 EUR
AATB10041120 AATB10041121
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (mFluor™ Violet 540)
Antibodies
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: mFluor™ Violet 540
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR