Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (APC/iFluor® 700)

Supplier: AAT BIOQUEST
100511F0 100511F1 100511F2
AATB100511F0EA 319 EUR
AATB100511F0 AATB100511F1 AATB100511F2
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (APC/iFluor® 700)
Antibodies
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD5 antigen
Clonality: Monoclonal
Clone: L17F12
Conjugation: APC/iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD5
Environmentally Preferable
Antigen name CD5 antigen
Antibody type Primary
Clonality Monoclonal
Conjugation APC (Allophycocyanin)/iFluor® 700
Clone L17F12
Reactivity Human
Host Mouse
Isotype
ImmunoFluorescence Yes
Flow cytometry Yes
Purification Affinity chromatography
Cross adsorption No
Storage temperature 2...8 °C with minimised light exposure. Do not freeze.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR