Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 514)

Supplier: AAT BIOQUEST
10041060 10041061
AATB10041060EA 371 EUR
AATB10041060 AATB10041061
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 514)
Antibodies
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD4 antigen
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: iFluor® 514
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR