Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 514)

Supplier: AAT BIOQUEST
10441060 10441061
AATB10441060EA 371 EUR
AATB10441060 AATB10441061
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 514)
Antibodies
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD44 antigen
Clonality: Monoclonal
Clone: IM7
Conjugation: iFluor® 514
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR