Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (mFluor™ Violet 510)
Supplier: AAT BIOQUEST
10141110 10141111
AATB10141110EA 371 EUR
AATB10141110 AATB10141111
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (mFluor™ Violet 510)
Antibodies
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD14 antigen
Clonality: Monoclonal
Clone: 61D3
Conjugation: mFluor™ Violet 510
Epitope:
Host: Mouse
Isotype:
Reactivity: Human