Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (mFluor™ Violet 500)
Supplier: AAT BIOQUEST
10021100 10021101
AATB10021100EA 371 EUR
AATB10021100 AATB10021101
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (mFluor™ Violet 500)
Antibodies
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: mFluor™ Violet 500
Epitope:
Host: Mouse
Isotype:
Reactivity: Human